ENA311758961 Enamine

1-(2-chloroacetyl)-N-(3-methylbutyl)piperidine-4-carboxamide

  • MDL number MFCD18089372
  • Cas Number : 1308650-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48984

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48984
Availability details for
Quantity
  • Availability
  • Add to Cart