ENA311758962 Enamine

N-[1-(2-chloroacetyl)piperidin-4-yl]-2-methylbenzamide

  • MDL number MFCD17976966
  • Cas Number : 1258640-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49045

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49045
Availability details for
Quantity
  • Availability
  • Add to Cart