ENA311758968 Enamine

2,2,2-trifluoroethyl N-(4-{imidazo[2,1-b][1,3]thiazol-6-yl}phenyl)carbamate

  • MDL number MFCD18089374
  • Cas Number : 1308650-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56616
Availability details for
Quantity
  • Availability
  • Add to Cart