ENA311758975 Enamine

N,N-dimethyl-5H,6H,7H,8H-pyrido[4,3-d]pyrimidin-4-amine

  • MDL number MFCD17976977
  • Cas Number : 1258640-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62881

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62881
Availability details for
Quantity
  • Availability
  • Add to Cart