ENA311758977 Enamine

3-{imidazo[1,2-a]pyridin-2-ylmethoxy}aniline dihydrochloride

  • MDL number MFCD17976981
  • Cas Number : 1258641-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65671

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65671
Availability details for
Quantity
  • Availability
  • Add to Cart