ENA311758983 Enamine

3-tert-butyl-4-methyl-1-phenyl-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD18089379
  • Cas Number : 1303889-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-68644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68644
Availability details for
Quantity
  • Availability
  • Add to Cart