ENA311758984 Enamine

4H,5H,6H,7H-[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD17976985
  • Cas Number : 1258649-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68658
Availability details for
Quantity
  • Availability
  • Add to Cart