ENA311758988 Enamine

3-(thiophen-3-yl)propan-1-amine hydrochloride

  • MDL number MFCD17976988
  • Cas Number : 1258651-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69815

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69815
Availability details for
Quantity
  • Availability
  • Add to Cart