ENA311758989 Enamine

4-(1,2,4-oxadiazol-3-yl)aniline hydrobromide

  • MDL number MFCD17976989
  • Cas Number : 1258651-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69825

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69825
Availability details for
Quantity
  • Availability
  • Add to Cart