ENA311758992 Enamine

4-(3,4-difluorophenyl)butan-2-amine hydrochloride

  • MDL number MFCD17976992
  • Cas Number : 1258652-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69892
Availability details for
Quantity
  • Availability
  • Add to Cart