ENA311758993 Enamine

5-methyl-4-sulfamoylthiophene-2-carboxylic acid

  • MDL number MFCD17976993
  • Cas Number : 1258639-67-6
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69908
Availability details for
Quantity
  • Availability
  • Add to Cart