ENA311758994 Enamine

2-(2-chloropyridin-4-yl)-5,6-dimethyl-1,4-dihydropyrimidin-4-one

  • MDL number MFCD26143907
  • Cas Number : 1258650-00-8
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69934
Availability details for
Quantity
  • Availability
  • Add to Cart