ENA311758999 Enamine

N1-{3-bromo-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl}ethane-1,2-diamine

  • MDL number MFCD17976999
  • Cas Number : 1258651-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69954
Availability details for
Quantity
  • Availability
  • Add to Cart