ENA311759001 Enamine

5-(aminomethyl)-4-(propan-2-yl)-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD17977001
  • Cas Number : 1258639-70-1
  • Molecular Weight : 208.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69973
Availability details for
Quantity
  • Availability
  • Add to Cart