ENA311759003 Enamine

2-(3-aminopropyl)-4-methyl-1,3-thiazole-5-carboxylic acid dihydrochloride

  • MDL number MFCD17977003
  • Cas Number : 1258650-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69976

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69976
Availability details for
Quantity
  • Availability
  • Add to Cart