ENA311759011 Enamine

6-bromo-8-chloro-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number MFCD17977013
  • Cas Number : 1258651-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-70016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70016
Availability details for
Quantity
  • Availability
  • Add to Cart