ENA311759013 Enamine

4-(pyridin-2-yl)butanoic acid hydrochloride

  • MDL number MFCD17977015
  • Cas Number : 638167-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70038
Availability details for
Quantity
  • Availability
  • Add to Cart