ENA311759024 Enamine

pyrrolidine-3-carboxamide hydrochloride

  • MDL number MFCD17977027
  • Cas Number : 644972-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70092

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-70092
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart