ENA311759031 Enamine

[(1-benzyl-4-methylpiperidin-4-yl)methyl](methyl)amine

  • MDL number MFCD17977035
  • Cas Number : 403513-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70249

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70249
Availability details for
Quantity
  • Availability
  • Add to Cart