ENA311759035 Enamine

2-[1-(2,2-diethoxyethyl)-1H-1,2,3-triazol-4-yl]ethan-1-ol

  • MDL number MFCD17977039
  • Cas Number : 915369-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70269
Availability details for
Quantity
  • Availability
  • Add to Cart