ENA311759036 Enamine

2-chloro-[1,3]oxazolo[5,4-c]pyridine hydrochloride

  • MDL number MFCD17977040
  • Cas Number : 1258650-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70270

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-70270
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart