ENA311759037 Enamine

2-(1,4-diazepan-1-yl)-N-(prop-2-yn-1-yl)acetamide dihydrochloride

  • MDL number MFCD17977041
  • Cas Number : 1258650-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-70271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70271
Availability details for
Quantity
  • Availability
  • Add to Cart