ENA311759043 Enamine

5-(chloromethyl)-3-(furan-3-yl)-1,2,4-oxadiazole

  • MDL number MFCD17977046
  • Cas Number : 1258651-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70330
Availability details for
Quantity
  • Availability
  • Add to Cart