ENA311759046 Enamine

1-(3,4-dichloro-2-hydroxyphenyl)propan-1-one

  • MDL number MFCD17977049
  • Cas Number : 777067-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70341
Availability details for
Quantity
  • Availability
  • Add to Cart