ENA311759050 Enamine

[(2,3,4-trimethoxyphenyl)methyl]hydrazine dihydrochloride

  • MDL number MFCD17977053
  • Cas Number : 1258650-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70392
Availability details for
Quantity
  • Availability
  • Add to Cart