ENA311759052 Enamine

3-{5H,6H,7H-pyrrolo[2,1-c][1,2,4]triazol-3-yl}piperidine hydrochloride

  • MDL number MFCD17977057
  • Cas Number : 1258651-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70448
Availability details for
Quantity
  • Availability
  • Add to Cart