ENA311759054 Enamine

1-(4-phenyl-1,3-thiazol-2-yl)ethan-1-ol

  • MDL number MFCD17977059
  • Cas Number : 41029-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70462
Availability details for
Quantity
  • Availability
  • Add to Cart