ENA311759057 Enamine

1-[4-(2-chlorophenyl)oxan-4-yl]methanamine hydrochloride

  • MDL number MFCD17977062
  • Cas Number : 1258650-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70473
Availability details for
Quantity
  • Availability
  • Add to Cart