ENA311759058 Enamine

3-methyl-3-phenylcyclobutan-1-amine hydrochloride

  • MDL number MFCD17977063
  • Cas Number : 1258639-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70477
Availability details for
Quantity
  • Availability
  • Add to Cart