ENA311759059 Enamine

methyl 4-(chlorosulfonyl)thiophene-2-carboxylate

  • MDL number MFCD17977064
  • Cas Number : 97272-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70483
Availability details for
Quantity
  • Availability
  • Add to Cart