ENA311759061 Enamine

1-(1-oxo-1-phenylpropan-2-yl)pyrrolidine-2,5-dione

  • MDL number MFCD17977066
  • Cas Number : 96679-45-7
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70491
Availability details for
Quantity
  • Availability
  • Add to Cart