ENA311759064 Enamine

6-acetyl-5-methyl-2,3-dihydro-1H-pyrrolizine-7-carbonitrile

  • MDL number MFCD17977070
  • Cas Number : 1258639-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70508

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70508
Availability details for
Quantity
  • Availability
  • Add to Cart