ENA311759071 Enamine

4-methyl-5-(piperidin-3-yl)-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD17977078
  • Cas Number : 1258651-55-6
  • Molecular Weight : 234.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70558
Availability details for
Quantity
  • Availability
  • Add to Cart