ENA311759072 Enamine

4-ethyl-5-(piperidin-3-yl)-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD17977079
  • Cas Number : 1258652-37-7
  • Molecular Weight : 248.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70559
Availability details for
Quantity
  • Availability
  • Add to Cart