ENA311759083 Enamine

4-(pyrrolidin-1-yl)benzene-1-carboximidamide hydrochloride

  • MDL number MFCD17977091
  • Cas Number : 1258650-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70604
Availability details for
Quantity
  • Availability
  • Add to Cart