ENA311759085 Enamine

ethyl N-(2-aminoethyl)carbamate hydrochloride

  • MDL number MFCD17977093
  • Cas Number : 1258639-97-2
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70623
Availability details for
Quantity
  • Availability
  • Add to Cart