ENA311759087 Enamine

methyl 4-amino-4-phenylbutanoate hydrochloride

  • MDL number MFCD17977095
  • Cas Number : 56523-55-8
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70627
Availability details for
Quantity
  • Availability
  • Add to Cart