ENA311759093 Enamine

3-cyclopropyl-1-(pyrimidin-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD17977102
  • Cas Number : 187795-41-1
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70680
Availability details for
Quantity
  • Availability
  • Add to Cart