ENA311759095 Enamine

1-(2-aminoethoxy)-2-(trifluoromethyl)benzene hydrochloride

  • MDL number MFCD17977105
  • Cas Number : 1258652-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70698

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70698
Availability details for
Quantity
  • Availability
  • Add to Cart