ENA311759097 Enamine

2-{2,4,6-trimethyl-3-oxo-1H,2H,3H-pyrazolo[3,4-b]pyridin-5-yl}acetic acid

  • MDL number MFCD17977109
  • Cas Number : 1258641-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70729
Availability details for
Quantity
  • Availability
  • Add to Cart