ENA311759098 Enamine

methyl 3-amino-2-[(2-methylphenyl)methyl]propanoate hydrochloride

  • MDL number MFCD17977111
  • Cas Number : 1258640-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70742
Availability details for
Quantity
  • Availability
  • Add to Cart