ENA311759100 Enamine

2-chloro-N-[(dimethyl-1,2-oxazol-4-yl)methyl]acetamide

  • MDL number MFCD17977113
  • Cas Number : 1258652-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70773
Availability details for
Quantity
  • Availability
  • Add to Cart