ENA311759102 Enamine

5,6-diethyl-3-oxo-2,3-dihydropyridazine-4-carbonitrile

  • MDL number MFCD17977115
  • Cas Number : 1258640-21-9
  • Molecular Weight : 177.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70787
Availability details for
Quantity
  • Availability
  • Add to Cart