ENA311759103 Enamine

2-(2,4-difluorophenoxy)ethan-1-amine hydrochloride

  • MDL number MFCD17977117
  • Cas Number : 263409-82-1
  • Molecular Weight : 209.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70798
Availability details for
Quantity
  • Availability
  • Add to Cart