ENA311759109 Enamine

methyl(2,2,3,3,3-pentafluoropropyl)amine hydrochloride

  • MDL number MFCD17977121
  • Cas Number : 2343-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70819

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70819
Availability details for
Quantity
  • Availability
  • Add to Cart