ENA311759112 Enamine

4-[(ethylamino)methyl]-3-fluorobenzonitrile hydrochloride

  • MDL number MFCD17977126
  • Cas Number : 1258652-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-71259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71259
Availability details for
Quantity
  • Availability
  • Add to Cart