ENA311759121 Enamine

4-(aminomethyl)-N-[2-(dimethylamino)ethyl]benzamide dihydrochloride

  • MDL number MFCD17977134
  • Cas Number : 1258640-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71307
Availability details for
Quantity
  • Availability
  • Add to Cart