ENA311759122 Enamine

N-[4-(aminomethyl)phenyl]-3-(pyrrolidin-1-yl)propanamide dihydrochloride

  • MDL number MFCD17977135
  • Cas Number : 1258650-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71312
Availability details for
Quantity
  • Availability
  • Add to Cart