ENA311759124 Enamine

methyl 1-(prop-2-yn-1-yl)piperidine-2-carboxylate

  • MDL number MFCD17977138
  • Cas Number : 1075220-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71377
Availability details for
Quantity
  • Availability
  • Add to Cart