ENA311759127 Enamine

2-(6-amino-9H-purin-9-yl)acetic acid dihydrochloride

  • MDL number MFCD17977141
  • Cas Number : 1258652-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71413
Availability details for
Quantity
  • Availability
  • Add to Cart